Learn More
Medchemexpress LLC Fbpase-1 inhibitor-1 | 883973-99-7 | 99.1% | 377.63 g/mol | C13H7Cl3N2O3S | 5 MG
SDP
Catalog No. 5000223330
Encompass_Preferred
FBPase-1 inhibitor-1 is an allosteric small-molecule inhibitor of fructose-1,6-bisphosphatase (FBPase-1) intended for laboratory research use. The compound has a benzoxazole benzenesulfonamide scaffold, molecular formula C13H7Cl3N2O3S, molecular weight 377.63 g/mol, and CAS 883973-99-7. It is supplied as solids in multiple sizes and as a 10 mM solution in DMSO for in vitro studies.
  • Allosteric inhibition of FBPase-1 for gluconeogenesis research.
  • High purity of 99.1% suitable for biochemical assays.
  • Available as solids in multiple sizes and as a DMSO solution.
  • Molecular formula C13H7Cl3N2O3S and molecular weight 377.63 g/mol for reference.
  • For research use only; not for human or clinical applications.
Catalog No. 50-002-23330 Supplier Medchemexpress LLC Supplier No. HY1367175MG
FBPase-1 inhibitor-1 is an allosteric small-molecule inhibitor of fructose-1,6-bisphosphatase (FBPase-1) intended for laboratory research use. The compound has a benzoxazole benzenesulfonamide scaffold, molecular formula C13H7Cl3N2O3S, molecular weight 377.63 g/mol, and CAS 883973-99-7. It is supplied as solids in multiple sizes and as a 10 mM solution in DMSO for in vitro studies.
  • Allosteric inhibition of FBPase-1 for gluconeogenesis research.
  • High purity of 99.1% suitable for biochemical assays.
  • Available as solids in multiple sizes and as a DMSO solution.
  • Molecular formula C13H7Cl3N2O3S and molecular weight 377.63 g/mol for reference.
  • For research use only; not for human or clinical applications.