Learn More
MSC Carlisle Fiberglass Handle, 48 Inch, 1 Inch Diameter, Red, Threaded
Catalog No. 502840001
Encompass
This 48 inch red threaded fiberglass handle is designed to keep floors clean. It features a durable fiberglass construction to withstand the demands of commercial environments such as restaurants, office buildings, or grocery stores. The threaded tip allows for easy installation with various compatible brooms (sold separately), and an innovative, self-locking flex-tip provides greater flexibility and reduces handle stress. The red color facilitates a color-coded cleaning system, aligning with HACCP plans to prevent cross-contamination and enhance food safety.
  • Helps with HACCP plans through color-coded design.
  • Durable fiberglass construction for demanding use.
  • Self-locking flex head reduces stress on handle.
  • Threaded tip allows for easy installation of compatible brooms.
  • Features a hanging hole for convenient storage.
Catalog No. 50-284-0001 Supplier MSC Supplier No. 95750865
This 48 inch red threaded fiberglass handle is designed to keep floors clean. It features a durable fiberglass construction to withstand the demands of commercial environments such as restaurants, office buildings, or grocery stores. The threaded tip allows for easy installation with various compatible brooms (sold separately), and an innovative, self-locking flex-tip provides greater flexibility and reduces handle stress. The red color facilitates a color-coded cleaning system, aligning with HACCP plans to prevent cross-contamination and enhance food safety.
  • Helps with HACCP plans through color-coded design.
  • Durable fiberglass construction for demanding use.
  • Self-locking flex head reduces stress on handle.
  • Threaded tip allows for easy installation of compatible brooms.
  • Features a hanging hole for convenient storage.