Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Fetal Bovine Serum (FBS), USDA Premium Grade, Heat Inactivated, 500 mL
SDP

Catalog No. 50983277
Encompass_Preferred

Commonly used as a supplement in classical cell culture media. Nourishes and protects the cells in a way that almost nothing else has been found to do.

Heat-inactivation of FBS is done under controlled conditions, heating at 56°C for 30 minutes. This treatment inactivates components in the complement system and other contaminants that may influence cell growth.

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 12 months

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-277 Supplier Quality Biological Inc Supplier No. 110001101HI
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Commonly used as a supplement in classical cell culture media. Nourishes and protects the cells in a way that almost nothing else has been found to do.

Heat-inactivation of FBS is done under controlled conditions, heating at 56°C for 30 minutes. This treatment inactivates components in the complement system and other contaminants that may influence cell growth.

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 12 months

Product and/or packaging contains recycled/renewable content Product and/or packaging is recyclable or reduces waste Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.