Learn More
Omega Scientific Inc Fetal Bovine Serum, Dialyzed, 500 Ml
Catalog No. NC1290080
Encompass_Preferred
FB-03: Fetal Bovine Serum, Dialyzed, 500 mLDialzyed Fetal Bovine Serum is passed through a 0.15M NaCl column to reduce the concentrations of low molecular weight components. The molecular weight cut off used in this process is 10,000.
Catalog No. NC1290080 Supplier Omega Scientific Inc Supplier No. FB03
FB-03: Fetal Bovine Serum, Dialyzed, 500 mLDialzyed Fetal Bovine Serum is passed through a 0.15M NaCl column to reduce the concentrations of low molecular weight components. The molecular weight cut off used in this process is 10,000.