Learn More
STEMCELL Technologies STEMdiff™ Endothelial Expansion Medium Kit, Size: 1 Kit
SDP
Catalog No. 502045467
Encompass
STEMdiff™ Endothelial Expansion Medium Kit (Catalog #08007) includes the xeno-free endothelial expansion basal medium and supplement, and must be used in conjunction with Animal Component-Free Cell Attachment Substrate. STEMdiff™ Endothelial Expansion Medium is optimized for the expansion of hPSC-derived endothelial cells and for the maintenance of endothelial marker expression. For passaging, Animal Component-Free Cell Dissociation Kit (Catalog #05426) is required.NOTE: Heparin Solution (Catalog #07980) is required for preparation of complete STEMdiff™ Endothelial Expansion Medium.CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.
Catalog No. 50-204-5467 Supplier STEMCELL Technologies Supplier No. 08007
STEMdiff™ Endothelial Expansion Medium Kit (Catalog #08007) includes the xeno-free endothelial expansion basal medium and supplement, and must be used in conjunction with Animal Component-Free Cell Attachment Substrate. STEMdiff™ Endothelial Expansion Medium is optimized for the expansion of hPSC-derived endothelial cells and for the maintenance of endothelial marker expression. For passaging, Animal Component-Free Cell Dissociation Kit (Catalog #05426) is required.NOTE: Heparin Solution (Catalog #07980) is required for preparation of complete STEMdiff™ Endothelial Expansion Medium.CollPlant (https://collplant.com/) is the manufacturer of the rhCollagen component of the cell attachment substrate.