Learn More
CAPTIVATE BIO LLC FBS Heat-Inactivated USDA
Catalog No. 5000876334
Encompass_Preferred
Captivate Bio s Premium Heath-Inactivated Fetal Bovine Serum USDA is processed and filtered under stringent quality control Packaged in 500mL bottle Serum is inactivated for 30 minutes at 56 C which inactivates viruses and destroys contaminants
Catalog No. 50-008-76334 Supplier CAPTIVATE BIO LLC Supplier No. FBS1620HA
Captivate Bio s Premium Heath-Inactivated Fetal Bovine Serum USDA is processed and filtered under stringent quality control Packaged in 500mL bottle Serum is inactivated for 30 minutes at 56 C which inactivates viruses and destroys contaminants