Learn More
CAPTIVATE BIO LLC FBS Heat-Inactivated USDA
Catalog No. 5000876337
Encompass_Preferred
Captivate Bio s Premium Heath-Inactivated Fetal Bovine Serum USDA is processed and filtered under stringent quality control Packaged in 6x500mL bottles Serum is inactivated for 30 minutes at 56 C which inactivates viruses and destroys contaminants
Catalog No. 50-008-76337 Supplier CAPTIVATE BIO LLC Supplier No. FBS1620HA6
Captivate Bio s Premium Heath-Inactivated Fetal Bovine Serum USDA is processed and filtered under stringent quality control Packaged in 6x500mL bottles Serum is inactivated for 30 minutes at 56 C which inactivates viruses and destroys contaminants