Learn More
VWR INTERNATIONAL Premium Grade Fetal Bovine Serum (FBS) | 500mL
Catalog No. NC1716838
Encompass_Preferred
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications. 100% US Origin. High quality, high performance FBS for a variety of applications. Preservation of native, growth-promoting factors and minimization of growth-inhibiting factors. ISIA Certified Supply Chain for Traceability.
Catalog No. NC1716838 Supplier VWR INTERNATIONAL Supplier No. 97068085
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications. 100% US Origin. High quality, high performance FBS for a variety of applications. Preservation of native, growth-promoting factors and minimization of growth-inhibiting factors. ISIA Certified Supply Chain for Traceability.