Learn More
CAPTIVATE BIO LLC FBS USDA Premium
Supplier: CAPTIVATE BIO LLC FBS16206
Captivate Bio s Premium Fetal Bovine Serum USDA is triple 0 1 m sterile filtered Sourced from USDA-approved abattoirs and processed and filtered under stringent quality control This product is available in packs 6x500mL bottles Ideal for general cell culture with common cell lines