Learn More
Aviva Systems Biology Corp FBXL10 Antibody - middle region (ARP39706_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39706_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXL10. Alternate names: CXXC2, Fbl10, JHDM1B, KDM2B, PCCX2, FBXL10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57145