Learn More
BPS Bioscience Inc FBXL10 (KDM2B) Homogeneous Assay Kit, 384 reactions
Supplier: BPS Bioscience Inc 506102
The FBXL10 (KDM2B) Homogenous Assay Kit is designed to measure activity of the FBXL10 (F-box and leucine-rich repeat protein 10) for screening and profiling purposes. The kit comes in a convenient format 96-well or 384-well AlphaLISA® format, with enough purified recombinant FBXL10, primary antibody, substrate and buffers for 100 or 400 enzyme reactions.