Learn More
Aviva Systems Biology Corp FBXL11 Antibody - N-terminal region (ARP31697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL11. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FBXL11. Alternate names: FBL7, CXXC8, FBL11, FBXL11, JHDM1A, LILINAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54310