Learn More
BPS Bioscience Inc FBXL11 (KDM2A) Homogeneous Assay Kit, 384 reactions
SDP
Catalog No. NC4036025
Encompass
The FBXL11 (KDM2A) Homogeneous Assay Kit is designed to measure FBXL11 (F-box and leucine-rich repeat protein 11) activity for screening and profiling applications. The FBXL11 (KDM2A) Homogeneous Assay Kit comes in a convenient 96-well or 384-well AlphaLISA® format, with enough purified recombinant FBXL11 (amino acids 2-700), substrate, primary antibody, assay and detection buffer for 100 or 400 enzyme reactions.
Catalog No. NC4036025 Supplier BPS Bioscience Inc Supplier No. 506112
The FBXL11 (KDM2A) Homogeneous Assay Kit is designed to measure FBXL11 (F-box and leucine-rich repeat protein 11) activity for screening and profiling applications. The FBXL11 (KDM2A) Homogeneous Assay Kit comes in a convenient 96-well or 384-well AlphaLISA® format, with enough purified recombinant FBXL11 (amino acids 2-700), substrate, primary antibody, assay and detection buffer for 100 or 400 enzyme reactions.