Learn More
Aviva Systems Biology Corp FBXL12 Antibody - C-terminal region (ARP59409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FBXL12. Alternate names: FLJ20188, Fbl12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-689