Learn More
ABclonal Technology FBXL12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14589500UL
Encompass_Preferred
Members of the F-box protein family, such as FBXL12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1, MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
Catalog No. 50-001-06351