Learn More
Aviva Systems Biology Corp FBXL14 Antibody - N-terminal region (ARP52968_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52968_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXL14. Alternate names: Fbl14, MGC40195ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60881