Learn More
Aviva Systems Biology Corp FBXL20 Antibody - middle region (ARP60708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL20. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FBXL20. Alternate names: FLJ21037, Fbl2, Fbl20, MGC15482, SCR, SCRAPPERApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-137