Learn More
Aviva Systems Biology Corp FBXL3 Antibody - middle region (ARP43098_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43098_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXL3. Alternate names: FBL3, FBL3A, FBXL3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-58429