Learn More
Aviva Systems Biology Corp Fbxl3 Antibody - C-terminal region (ARP43099_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43099_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fbxl3. It was validated on Western Blot. . Alternate names: AU041772, AW212966, FBK, Fbl3a, Fbxl3a, OvtmApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-053