Learn More
Aviva Systems Biology Corp FBXL4 Antibody - N-terminal region (ARP54856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FBXL4. Alternate names: FBL4, FBL5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-972