Learn More
Aviva Systems Biology Corp FBXL5 Antibody - middle region (ARP43105_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43105_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FBXL5. Alternate names: FBL4, FBL5, FLR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-054