Learn More
Aviva Systems Biology Corp FBXL8 Antibody - N-terminal region (ARP62815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXL8. It was validated on Western Blot. . Alternate names: FBL8, FLJ11278, MGC19959ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-776