Learn More
Aviva Systems Biology Corp Fbxo11 Antibody - C-terminal region (ARP43326_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43326_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fbxo11. It was validated on Western Blot. . Alternate names: C80048, Jf, MGC156416ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-067