Learn More
Aviva Systems Biology Corp FBXO16 Antibody - N-terminal region (ARP53226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXO16. Alternate names: FBX16, MGC125923, MGC125924, MGC125925ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60978