Learn More
Aviva Systems Biology Corp FBXO21 Antibody - N-terminal region (ARP43175_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43175_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXO21. Alternate names: DKFZp434G058, FBX21, FLJ90233, KIAA0875, MGC26682ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-58456