Learn More
Aviva Systems Biology Corp FBXO24 Antibody - middle region (ARP43378_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43378_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO24. Alternate names: DKFZp434I1122, FBX24ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58513