Learn More
Aviva Systems Biology Corp FBXO25 Antibody - N-terminal region (ARP43117_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43117_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXO25. Alternate names: FBX25, MGC20256, MGC51975ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58434