Learn More
Aviva Systems Biology Corp FBXO28 Antibody - middle region (ARP55191_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55191_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO28. Alternate names: FLJ10766, Fbx28, KIAA0483, CENP-30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61436