Learn More
Aviva Systems Biology Corp FBXO3 Antibody - N-terminal region (ARP43119_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43119_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXO3. Alternate names: DKFZp564B092, FBA, FBX3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58436