Learn More
Aviva Systems Biology Corp FBXO32 Antibody - N-terminal region (ARP52865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO32. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FBXO32. Alternate names: FLJ32424, Fbx32, MAFbx, MGC33610ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-778