Learn More
Aviva Systems Biology Corp Fbxo32 Antibody - C-terminal region (ARP52866_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52866_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fbxo32. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region from the human protein.. Alternate names: 4833442G10Rik, AI430017, ATROGIN1, MAFbx, atrogin-1, Gm20361ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-779