Learn More
Aviva Systems Biology Corp FBXO33 Antibody - middle region (ARP55943_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO33. Alternate names: Fbx33, c14_5247, BMND12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61670