Learn More
Aviva Systems Biology Corp Fbxo34 Antibody - N-terminal region (ARP57070_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57070_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fbxo34. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2900057B08Rik, 5830426G16RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-263