Learn More
Aviva Systems Biology Corp FBXO34 Antibody - middle region (ARP57071_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57071_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO34. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO34. Alternate names: CGI-301, DKFZp547C162, FLJ20725, Fbx34, MGC126434, MGC126435ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53944