Learn More
Aviva Systems Biology Corp FBXO42 Antibody - middle region (ARP47968_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47968_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO42. Alternate names: Fbx42, KIAA1332, JFKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59689