Learn More
Aviva Systems Biology Corp FBXO7 Antibody - middle region (ARP43128_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43128_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO7. Alternate names: DKFZp686B08113, FBX, FBX07, FBX7, PKPS, PARK15ApplicationIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58441