Learn More
Aviva Systems Biology Corp FBXW11 Antibody - N-terminal region (ARP43379_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43379_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXW11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBXW11. Alternate names: BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos, KIAA0696ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58514