Learn More
Aviva Systems Biology Corp Fbxw7 Antibody - middle region (ARP37443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fbxw7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Fbxw7. Alternate names: 1110001A17Rik, AGO, Cdc4, Fbw7, Fbwd6, Fbx30, Fbxo30, Fbxw6, SEL-10ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-633