Learn More
BD Cell Analysis 3P BD™ FC Beads BV421, Flow Cytometry Compensation, Instrument Standardization, 15 pcs
Catalog No. NC1562741
Encompass
BD FC Beads BV421 are specialized fluorescent beads designed for flow cytometry compensation and instrument standardization. They ensure accurate spillover values and consistent data acquisition across multiple BD flow cytometers, enhancing experimental reliability. The beads are stable at 2°C–8°C and should be stored in a foil pouch, protected from light and humidity.
Catalog No. NC1562741 Supplier BD Cell Analysis 3P Supplier No. 661627
BD FC Beads BV421 are specialized fluorescent beads designed for flow cytometry compensation and instrument standardization. They ensure accurate spillover values and consistent data acquisition across multiple BD flow cytometers, enhancing experimental reliability. The beads are stable at 2°C–8°C and should be stored in a foil pouch, protected from light and humidity.