Learn More
BD Cell Analysis 3P FC Beads BV605
Catalog No. NC1745188
Encompass
BD™ FC beads are fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers. The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument. BD FC beads can also be used to standardize multiple instruments.
Catalog No. NC1745188 Supplier BD Cell Analysis 3P Supplier No. 661626
BD™ FC beads are fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers. The combination of BD™ CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument. BD FC beads can also be used to standardize multiple instruments.