Learn More
BD Cell Analysis 3P BD™ FC Beads BV711, Accurate Compensation, Instrument Standardization, 5 Tests
Catalog No. NC1880874
Encompass
The BD™ FC Beads BV711 facilitate precise fluorescence compensation in flow cytometry, enhancing accuracy and consistency across instruments. These fluorescent beads support the standardization of multiple flow cytometers, enabling reliable experimental results. Designed for stability, they ensure optimal performance when stored correctly, making them a valuable tool for researchers in need of dependable flow cytometry solutions.
Catalog No. NC1880874 Supplier BD Cell Analysis 3P Supplier No. 661629
The BD™ FC Beads BV711 facilitate precise fluorescence compensation in flow cytometry, enhancing accuracy and consistency across instruments. These fluorescent beads support the standardization of multiple flow cytometers, enabling reliable experimental results. Designed for stability, they ensure optimal performance when stored correctly, making them a valuable tool for researchers in need of dependable flow cytometry solutions.