Learn More
BD Cell Analysis 3P FC BEADS BV786 5 TESTS
Catalog No. NC1655628
Encompass
fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers
Catalog No. NC1655628 Supplier BD Cell Analysis 3P Supplier No. 661630
fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD™ flow cytometers