Learn More
BD Cell Analysis 3P BD FC Beads V450, For Flow Cytometry Calibration, Consistent Results, 5 Tests
Catalog No. NC1616925
Encompass
The BD™ FC Beads V450 are fluorescent beads designed for optimizing flow cytometry applications. These beads enable accurate calibration and standardization, ensuring consistent spillover values and a reliable fluorescence compensation matrix. When used in conjunction with BD CS&T beads, they enhance the voltage and compensation settings across all instrument channels, making them ideal for researchers seeking reproducible results. Designed for performance, these beads are a crucial tool in scientific research workflows.
Catalog No. NC1616925 Supplier BD Cell Analysis 3P Supplier No. 661623
The BD™ FC Beads V450 are fluorescent beads designed for optimizing flow cytometry applications. These beads enable accurate calibration and standardization, ensuring consistent spillover values and a reliable fluorescence compensation matrix. When used in conjunction with BD CS&T beads, they enhance the voltage and compensation settings across all instrument channels, making them ideal for researchers seeking reproducible results. Designed for performance, these beads are a crucial tool in scientific research workflows.