Learn More
BD Cell Analysis 3P Flourescent Beads, FC Beads V500-C, 5 Tests
Catalog No. NC1562742
Encompass
BD FC beads are fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD flow cytometers. The combination of BD CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument. BD FC beads can also be used to standardize multiple instruments.
Catalog No. NC1562742 Supplier BD Cell Analysis 3P Supplier No. 661624
BD FC beads are fluorescent beads that enable the software to determine spillover values (SOVs) for fluorescence compensation and calculate a fluorescence compensation matrix during setup of BD flow cytometers. The combination of BD CS&T beads and BD FC beads accurately sets the voltage and compensation for each of the channels of the instrument. BD FC beads can also be used to standardize multiple instruments.