Learn More
Medchemexpress LLC Fc epsilon RIA/FCER1 10ug
SDP
Supplier:  Medchemexpress LLC HYP7219110UG
Encompass_Preferred
Recombinant human Fc epsilon RIA (FCER1A) is the extracellular alpha chain of the high-affinity IgE receptor supplied as a His-SUMO-tagged recombinant protein. The lyophilized protein is intended for use in binding studies, ELISA, and other immunological or biochemical assays requiring the human FCER1A ectodomain.
  • His-SUMO tagged protein for affinity purification and detection.
  • Expressed in E. coli and supplied lyophilized for enhanced stability.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Formulated with trehalose and EDTA for increased storage stability.
  • Suitable for ELISA and binding assays; functional activity characterized.
  • Recommended reconstitution at or above 100 μg/mL for optimal performance.
Catalog No. 50-001-83548