Learn More
Medchemexpress LLC Fc epsilon RIA/FCER1 50ug
SDP
Catalog No. 5000183292
Encompass_Preferred
Recombinant human Fc epsilon RI alpha (extracellular domain) supplied as 50 μg lyophilized material with an N-terminal His-SUMO tag, intended for binding and functional studies involving human IgE.
  • Prepared as lyophilized powder for long-term storage and transport.
  • Includes N-terminal His-SUMO tag to facilitate affinity purification.
  • Expression system: E. coli for efficient production.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~37 kDa for molecular characterization.
  • Demonstrated IgE binding in ELISA with reported ED50 of 5.864 μg/mL.
Catalog No. 50-001-83292 Supplier Medchemexpress LLC Supplier No. HYP7219150UG
Recombinant human Fc epsilon RI alpha (extracellular domain) supplied as 50 μg lyophilized material with an N-terminal His-SUMO tag, intended for binding and functional studies involving human IgE.
  • Prepared as lyophilized powder for long-term storage and transport.
  • Includes N-terminal His-SUMO tag to facilitate affinity purification.
  • Expression system: E. coli for efficient production.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~37 kDa for molecular characterization.
  • Demonstrated IgE binding in ELISA with reported ED50 of 5.864 μg/mL.