Learn More
Medchemexpress LLC Fc epsilon RIA/FCER1A protein, mouse (HEK293, C-hFc) | >85.0% | 5 UG
SDP
Catalog No. 5000292861
Encompass_Preferred
Recombinant mouse Fc epsilon RI alpha (FCER1A) extracellular domain fused to a C-terminal human Fc, expressed in HEK293 cells and supplied lyophilized. Intended for immunoassays, binding studies, and functional assays involving IgE-FcεRI interactions; documentation including datasheet and COA is available.
Catalog No. 50-002-92861 Supplier Medchemexpress LLC Supplier No. HYP752125UG
Recombinant mouse Fc epsilon RI alpha (FCER1A) extracellular domain fused to a C-terminal human Fc, expressed in HEK293 cells and supplied lyophilized. Intended for immunoassays, binding studies, and functional assays involving IgE-FcεRI interactions; documentation including datasheet and COA is available.