Learn More
Medchemexpress LLC Fc gamma receptor IIa (CD32a) protein, rhesus macaque | >95.0% | 5 UG
SDP
Catalog No. 5000292524
Encompass_Preferred
Recombinant Fc gamma receptor IIa (CD32a) protein from rhesus macaque, expressed in HEK293 cells with a C-terminal His tag. The protein binds the Fc region of IgG and functions as a low-affinity Fcγ receptor involved in immune regulation and downstream signaling.
Catalog No. 50-002-92524 Supplier Medchemexpress LLC Supplier No. HYP756475UG
Recombinant Fc gamma receptor IIa (CD32a) protein from rhesus macaque, expressed in HEK293 cells with a C-terminal His tag. The protein binds the Fc region of IgG and functions as a low-affinity Fcγ receptor involved in immune regulation and downstream signaling.