Learn More
Genscript Corporation Fc gamma RI/CD64Biotin His
Catalog No. 5000059519
Encompass
The Fc gamma Rs have been divided into three classes based on close relationships in their extracellular domains these groups are designated Fc gamma RI (also known as CD64) Fc gamma RII (CD32) and Fc gamma RIII (CD16) Each group may be encoded by multiple genes and exist in different isoforms depending on species and cell type
Catalog No. 50-000-59519 Supplier Genscript Corporation Supplier No. Z04449100
The Fc gamma Rs have been divided into three classes based on close relationships in their extracellular domains these groups are designated Fc gamma RI (also known as CD64) Fc gamma RII (CD32) and Fc gamma RIII (CD16) Each group may be encoded by multiple genes and exist in different isoforms depending on species and cell type