Learn More
Medchemexpress LLC Fc gamma RIIA/CD32a 100ug | 100ug
SDP
Catalog No. 5000187806
Encompass_Preferred
Fc gamma The RIIA/CD32a protein plays a key role as a low-affinity receptor that specifically binds to the Fc region of immunoglobulin gamma It interacts with IgG to initiate cellular responses against pathogens which is critical for immune regulation Fc gamma RIIA/CD32a Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIA/CD32a protein expressed by HEK293 with C-Avi C-His labeled tag and H167 mutation
Catalog No. 50-001-87806 Supplier Medchemexpress LLC Supplier No. HYP78100100UG
Fc gamma The RIIA/CD32a protein plays a key role as a low-affinity receptor that specifically binds to the Fc region of immunoglobulin gamma It interacts with IgG to initiate cellular responses against pathogens which is critical for immune regulation Fc gamma RIIA/CD32a Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Fc gamma RIIA/CD32a protein expressed by HEK293 with C-Avi C-His labeled tag and H167 mutation